Anti-PRKCQ Rabbit Polyclonal Antibody

Lieferant: Abcam
AB131503-100
ABCAAB131503-100EA 910 EUR
ABCAAB131503-100
Anti-PRKCQ Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRKCQ (protein kinase C, theta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR