Anti-EIF4E Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB131513-100EA 859 EUR
ABCAAB131513-100
Anti-EIF4E Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EIF4E (eukaryotic translation initiation factor 4E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR