Anti-FLT3 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB131545-100EA 840 EUR
ABCAAB131545-100
Anti-FLT3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: FLT3 (fms-related tyrosine kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human