Anti-PAK1 + PAK2 + PAK3 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB131547-100EA 835 EUR
ABCAAB131547-100
Anti-PAK1 + PAK2 + PAK3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAK1 + PAK2 + PAK3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat