Anti-Kappa light chain Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: TB28-2]
Lieferant: Abcam
ABCAAB131602-100EA 835 EUR
ABCAAB131602-100
Anti-Kappa light chain Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: TB28-2]
Antikörper
Type: Primary
Antigen: Kappa light chain
Clonality: monoclonal
Clone: TB28-2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human