Anti-SENP2 Mouse monoclonal antibody [clone: 1H2]

Lieferant: Abcam
ABCAAB131637-100EA 755 EUR
ABCAAB131637-100
Anti-SENP2 Mouse monoclonal antibody [clone: 1H2]
Type: Primary
Antigen: SENP2 (SUMO1/sentrin/SMT3 specific peptidase 2)
Clonality: monoclonal
Clone: 1H2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR