Anti-S Tag Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Abcam
ABCAAB133157-100EA 834 EUR
ABCAAB133157-100
Anti-S Tag Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Primary
Antigen: S Tag (S tag)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR