Anti-ITGA3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB133198-50EA 840 EUR
ABCAAB133198-50
Anti-ITGA3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ITGA3 (integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR