Anti-SSR1 Rabbit Monoclonal Antibody [clone: EPR5603]

Lieferant: Abcam
AB133238-100
ABCAAB133238-100EA 827 EUR
ABCAAB133238-100
Anti-SSR1 Rabbit Monoclonal Antibody [clone: EPR5603]
Antikörper
Type: Primary
Antigen: SSR1 (signal sequence receptor, alpha)
Clonality: monoclonal
Clone: EPR5603
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR