Anti-TAF9B Rabbit Monoclonal Antibody [clone: EPR5266]

Lieferant: Abcam
ABCAAB133239-100EA 758 EUR
ABCAAB133239-100
Anti-TAF9B Rabbit Monoclonal Antibody [clone: EPR5266]
Antikörper
Type: Primary
Antigen: TAF9B (TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31kDa)
Clonality: monoclonal
Clone: EPR5266
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR