Anti-NAPSIN A Rabbit Monoclonal Antibody [clone: EPR6252]

Lieferant: Abcam
ABCAAB133249-100EA 866 EUR
ABCAAB133249-100
Anti-NAPSIN A Rabbit Monoclonal Antibody [clone: EPR6252]
Antikörper
Type: Primary
Antigen: NAPSIN A
Clonality: monoclonal
Clone: EPR6252
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR