Anti-SMARCA4 Rabbit Monoclonal Antibody [clone: EPNCIR111B]
Lieferant: Abcam
ABCAAB133257-100EA 912 EUR
ABCAAB133257-100
Anti-SMARCA4 Rabbit Monoclonal Antibody [clone: EPNCIR111B]
Antikörper
Type: Primary
Antigen: SMARCA4 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4)
Clonality: monoclonal
Clone: EPNCIR111B
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen