Anti-RUNX1 / AML1 Rabbit Monoclonal Antibody [clone: EPR3098]

Lieferant: Abcam
AB133261-100
ABCAAB133261-100EA 898 EUR
ABCAAB133261-100
Anti-RUNX1 / AML1 Rabbit Monoclonal Antibody [clone: EPR3098]
Antikörper
Type: Primary
Antigen: RUNX1 / AML1
Clonality: monoclonal
Clone: EPR3098
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR