Anti-KRT18 Rabbit Monoclonal Antibody [clone: EPR1626]

Lieferant: Abcam
ABCAAB133263-100EA 949 EUR
ABCAAB133263-100
Anti-KRT18 Rabbit Monoclonal Antibody [clone: EPR1626]
Antikörper
Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: monoclonal
Clone: EPR1626
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR