Anti-ERG Rabbit Monoclonal Antibody [clone: EPR3864]

Lieferant: Abcam
ABCAAB133264-100EA 880 EUR
ABCAAB133264-100
Anti-ERG Rabbit Monoclonal Antibody [clone: EPR3864]
Antikörper
Type: Primary
Antigen: ERG (v-ets avian erythroblastosis virus E26 oncogene homolog)
Clonality: monoclonal
Clone: EPR3864
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR