Anti-INS Rabbit Monoclonal Antibody [clone: EPR3074]
Lieferant: Abcam
ABCAAB133281-100EA 835 EUR
ABCAAB133281-100
Anti-INS Rabbit Monoclonal Antibody [clone: EPR3074]
Antikörper
Type: Primary
Antigen: INS (insulin)
Clonality: monoclonal
Clone: EPR3074
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen