Anti-CR1 Rabbit Monoclonal Antibody [clone: EPR6601]

Lieferant: Abcam
ABCAAB133293-100EA 859 EUR
ABCAAB133293-100
Anti-CR1 Rabbit Monoclonal Antibody [clone: EPR6601]
Antikörper
Type: Primary
Antigen: CR1 (complement component (3b/4b) receptor 1 (Knops blood group))
Clonality: monoclonal
Clone: EPR6601
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR