Anti-SAA4 Rabbit Monoclonal Antibody [clone: EPR2927]

Lieferant: Abcam
ABCAAB133312-100EA 778 EUR
ABCAAB133312-100
Anti-SAA4 Rabbit Monoclonal Antibody [clone: EPR2927]
Antikörper
Type: Primary
Antigen: SAA4 (serum amyloid A4, constitutive)
Clonality: monoclonal
Clone: EPR2927
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR