Anti-ISG15 Rabbit Monoclonal Antibody [clone: EPR3446]

Lieferant: Abcam
ABCAAB133346-100EA 928 EUR
ABCAAB133346-100
Anti-ISG15 Rabbit Monoclonal Antibody [clone: EPR3446]
Antikörper
Type: Primary
Antigen: ISG15 (ISG15 ubiquitin-like modifier)
Clonality: monoclonal
Clone: EPR3446
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR