Anti-ANAPC1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB133397-50EA 866 EUR
ABCAAB133397-50
Anti-ANAPC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ANAPC1 (anaphase promoting complex subunit 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR