Anti-IgG1 + IgG2A + IgG3 Rabbit Monoclonal Antibody

Lieferant: Abcam
AB133469-100
ABCAAB133469-100EA 944 EUR
ABCAAB133469-100
Anti-IgG1 + IgG2A + IgG3 Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: IgG1 + IgG2A + IgG3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR