Anti-S100 ALPH Rabbit Monoclonal Antibody

Lieferant: Abcam
AB133519-100
ABCAAB133519-100EA 903 EUR
ABCAAB133519-100
Anti-S100 ALPH Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: S100 ALPH
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR