Anti-DLD Rabbit Monoclonal Antibody
Lieferant: Abcam
ABCAAB133551-100EA 779 EUR
ABCAAB133551-100
Anti-DLD Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: DLD
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen