Anti-CD36 Rabbit Monoclonal Antibody
Lieferant: Abcam
ABCAAB133625-100EA 980 EUR
ABCAAB133625-100
Anti-CD36 Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen