Anti-CASPR Rabbit Monoclonal Antibody
Lieferant: Abcam
ABCAAB133634-100EA 866 EUR
ABCAAB133634-100
Anti-CASPR Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CASPR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen