Anti-DSG1 Rabbit Monoclonal Antibody
Lieferant: Abcam
ABCAAB133662-100EA 835 EUR
ABCAAB133662-100
Anti-DSG1 Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: DSG1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen