Anti-NDRG3 Rabbit Monoclonal Antibody [clone: EPR9011]
Lieferant: Abcam
ABCAAB133715-100EA 780 EUR
ABCAAB133715-100
Anti-NDRG3 Rabbit Monoclonal Antibody [clone: EPR9011]
Antikörper
Type: Primary
Antigen: NDRG3 (NDRG family member 3)
Clonality: monoclonal
Clone: EPR9011
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen