Anti-MARK2 Rabbit Monoclonal Antibody [clone: EPR8553]

Lieferant: Abcam
ABCAAB133724-100EA 924 EUR
ABCAAB133724-100
Anti-MARK2 Rabbit Monoclonal Antibody [clone: EPR8553]
Antikörper
Type: Primary
Antigen: MARK2 (MAP/microtubule affinity-regulating kinase 2)
Clonality: monoclonal
Clone: EPR8553
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR