Anti-PTBP1 Rabbit Monoclonal Antibody [clone: EPR9048]

Lieferant: Abcam
AB133734-100
ABCAAB133734-100EA 898 EUR
ABCAAB133734-100
Anti-PTBP1 Rabbit Monoclonal Antibody [clone: EPR9048]
Antikörper
Type: Primary
Antigen: PTBP1 (polypyrimidine tract binding protein 1)
Clonality: monoclonal
Clone: EPR9048
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR