Anti-CLPTM1 Rabbit Monoclonal Antibody [clone: EPR8800]

Lieferant: Abcam
ABCAAB133756-100EA 785 EUR
ABCAAB133756-100
Anti-CLPTM1 Rabbit Monoclonal Antibody [clone: EPR8800]
Antikörper
Type: Primary
Antigen: CLPTM1 (cleft lip and palate associated transmembrane protein 1)
Clonality: monoclonal
Clone: EPR8800
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR