Anti-ARFGAP2 Rabbit Monoclonal Antibody [clone: EPR8497]

Lieferant: Abcam
ABCAAB133768-100EA 765 EUR
ABCAAB133768-100
Anti-ARFGAP2 Rabbit Monoclonal Antibody [clone: EPR8497]
Antikörper
Type: Primary
Antigen: ARFGAP2 (ADP-ribosylation factor GTPase activating protein 2)
Clonality: monoclonal
Clone: EPR8497
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR