Anti-TMED10 Rabbit Monoclonal Antibody [clone: EPR9037]

Lieferant: Abcam
ABCAAB133771-100EA 780 EUR
ABCAAB133771-100
Anti-TMED10 Rabbit Monoclonal Antibody [clone: EPR9037]
Antikörper
Type: Primary
Antigen: TMED10 (transmembrane emp24-like trafficking protein 10 (yeast))
Clonality: monoclonal
Clone: EPR9037
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR