Anti-Placental lactogen Rabbit Monoclonal Antibody [clone: EPR8264]

Lieferant: Abcam
ABCAAB133772-100EA 778 EUR
ABCAAB133772-100
Anti-Placental lactogen Rabbit Monoclonal Antibody [clone: EPR8264]
Antikörper
Type: Primary
Antigen: Placental lactogen
Clonality: monoclonal
Clone: EPR8264
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR