Anti-MRPL11 Rabbit Monoclonal Antibody [clone: EPR9111]

Lieferant: Abcam
ABCAAB133789-100EA 888 EUR
ABCAAB133789-100
Anti-MRPL11 Rabbit Monoclonal Antibody [clone: EPR9111]
Antikörper
Type: Primary
Antigen: MRPL11 (mitochondrial ribosomal protein L11)
Clonality: monoclonal
Clone: EPR9111
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR