Anti-APP Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB134022-50EA 840 EUR
ABCAAB134022-50
Anti-APP Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Chicken
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR