Anti-SLAMF1 Rabbit Monoclonal Antibody [clone: EPR5470]

Lieferant: Abcam
ABCAAB134044-100EA 892 EUR
ABCAAB134044-100
Anti-SLAMF1 Rabbit Monoclonal Antibody [clone: EPR5470]
Antikörper
Type: Primary
Antigen: SLAMF1 (signaling lymphocytic activation molecule family member 1)
Clonality: monoclonal
Clone: EPR5470
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR