Anti-CD63 Rabbit Monoclonal Antibody [clone: EPR5702]

Lieferant: Abcam
AB134045-100
ABCAAB134045-100EA 1020 EUR
ABCAAB134045-100
Anti-CD63 Rabbit Monoclonal Antibody [clone: EPR5702]
Antikörper
Type: Primary
Antigen: CD63 (CD63 molecule)
Clonality: monoclonal
Clone: EPR5702
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR