Anti-FOXP1 Rabbit Monoclonal Antibody [clone: EPR4113]

Lieferant: Abcam
AB134055-100
ABCAAB134055-100EA 972 EUR
ABCAAB134055-100
Anti-FOXP1 Rabbit Monoclonal Antibody [clone: EPR4113]
Antikörper
Type: Primary
Antigen: FOXP1 (forkhead box P1)
Clonality: monoclonal
Clone: EPR4113
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR