Anti-CEACAM6 Rabbit monoclonal antibody unconjugated [clone: EPR4404]

Lieferant: Abcam
ABCAAB134058-100EA 833 EUR
ABCAAB134058-100
Anti-CEACAM6 Rabbit monoclonal antibody unconjugated [clone: EPR4404]
Antikörper
Type: Primary
Antigen: CEACAM6 (carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen))
Clonality: monoclonal
Clone: EPR4404
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR