Anti-BST2 Rabbit Monoclonal Antibody [clone: EPR5648]

Lieferant: Abcam
AB134061-100
ABCAAB134061-100EA 903 EUR
ABCAAB134061-100
Anti-BST2 Rabbit Monoclonal Antibody [clone: EPR5648]
Antikörper
Type: Primary
Antigen: BST2 (bone marrow stromal cell antigen 2)
Clonality: monoclonal
Clone: EPR5648
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR