Anti-MCAM Rabbit Monoclonal Antibody [clone: EPR3207]

Lieferant: Abcam
ABCAAB134065-100EA 842 EUR
ABCAAB134065-100
Anti-MCAM Rabbit Monoclonal Antibody [clone: EPR3207]
Antikörper
Type: Primary
Antigen: MCAM (melanoma cell adhesion molecule)
Clonality: monoclonal
Clone: EPR3207
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR