Anti-CD93 Rabbit Monoclonal Antibody [clone: EPR5386]

Lieferant: Abcam
ABCAAB134079-100EA 777 EUR
ABCAAB134079-100
Anti-CD93 Rabbit Monoclonal Antibody [clone: EPR5386]
Antikörper
Type: Primary
Antigen: CD93 (CD93 molecule)
Clonality: monoclonal
Clone: EPR5386
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR