Anti-LSP1 Rabbit Monoclonal Antibody [clone: EPR3160]

Lieferant: Abcam
ABCAAB134099-100EA 778 EUR
ABCAAB134099-100
Anti-LSP1 Rabbit Monoclonal Antibody [clone: EPR3160]
Antikörper
Type: Primary
Antigen: LSP1 (lymphocyte-specific protein 1)
Clonality: monoclonal
Clone: EPR3160
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR