Anti-CD97 Rabbit Monoclonal Antibody [clone: EPR4428]
Lieferant: Abcam
ABCAAB134105-100EA 893 EUR
ABCAAB134105-100
Anti-CD97 Rabbit Monoclonal Antibody [clone: EPR4428]
Antikörper
Type: Primary
Antigen: CD97 (CD97 molecule)
Clonality: monoclonal
Clone: EPR4428
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen