Anti-Niemann Pick C1 Rabbit Monoclonal Antibody [clone: EPR5209]

Lieferant: Abcam
ABCAAB134113-100EA 928 EUR
ABCAAB134113-100
Anti-Niemann Pick C1 Rabbit Monoclonal Antibody [clone: EPR5209]
Antikörper
Type: Primary
Antigen: Niemann Pick C1
Clonality: monoclonal
Clone: EPR5209
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR