Anti-MUC2 Rabbit Monoclonal Antibody [clone: EPR6145]

Lieferant: Abcam
AB134119-100
ABCAAB134119-100EA 980 EUR
ABCAAB134119-100
Anti-MUC2 Rabbit Monoclonal Antibody [clone: EPR6145]
Antikörper
Type: Primary
Antigen: MUC2 (mucin 2, oligomeric mucus/gel-forming)
Clonality: monoclonal
Clone: EPR6145
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR