Anti-CD80 Rabbit Monoclonal Antibody [clone: EPR1157]

Lieferant: Abcam
ABCAAB134120-100EA 937 EUR
ABCAAB134120-100
Anti-CD80 Rabbit Monoclonal Antibody [clone: EPR1157]
Antikörper
Type: Primary
Antigen: CD80 (CD80 molecule)
Clonality: monoclonal
Clone: EPR1157
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR