Anti-FOS Rabbit monoclonal antibody unconjugated [clone: EPR883]

Lieferant: Abcam
ABCAAB134122-100EA 899 EUR
ABCAAB134122-100
Anti-FOS Rabbit monoclonal antibody unconjugated [clone: EPR883]
Antikörper
Type: Primary
Antigen: FOS (FBJ murine osteosarcoma viral oncogene homolog)
Clonality: monoclonal
Clone: EPR883
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR