Anti-CHD3 Rabbit Monoclonal Antibody [clone: EPNCIR110]

Lieferant: Abcam
ABCAAB134125-100EA 859 EUR
ABCAAB134125-100
Anti-CHD3 Rabbit Monoclonal Antibody [clone: EPNCIR110]
Antikörper
Type: Primary
Antigen: CHD3 (chromodomain helicase DNA binding protein 3)
Clonality: monoclonal
Clone: EPNCIR110
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR