Anti-ABL2 Rabbit Monoclonal Antibody
Lieferant: Abcam
ABCAAB134134-100EA 780 EUR
ABCAAB134134-100
Anti-ABL2 Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ABL2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen